2-Cyano-6-methoxybenzothiazole ≥97.0%

Supplier: TCI America

Synonyms: 6-Methoxybenzo[d]thiazole-2-carbonitrile6-Methoxy-2-benzothiazolecarbonitrile

C1176-1G
TCC1176-001GEA 108.3 USD
TCC1176-001G
2-Cyano-6-methoxybenzothiazole ≥97.0%
2-Cyano-6-methoxybenzothiazole

CAS Number: 943-03-3
MDL Number: MFCD00010537
Molecular Formula: C9H6N2OS
Molecular Weight: 190.22
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 130


Other Product Names / Synonyms: 6-Methoxy-2-benzothiazolecarbonitrile

Formula: C₉H₆N₂OS
MW: 190.22 g/mol
Melting Pt: 130 °C
MDL Number: MFCD00010537
CAS Number: 943-03-3
https://us.vwr.com/stibo/web/std.lang.all/84/28/28938428.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR