Butyl-2-chloroethyl ether ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloroethyl Butyl Ether1-(2-Chloroethoxy)butane

C1171-25ML
TCC1171-025MLEA 175.55 USD
TCC1171-025ML
Butyl-2-chloroethyl ether ≥98.0%
Butyl-2-chloroethyl ether

CAS Number: 10503-96-5
MDL Number: MFCD00059051
Molecular Formula: C6H13ClO
Molecular Weight: 136.62
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 50
Flash Point (°C): 46
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: 2-Chloroethyl Butyl Ether

Formula: C₆H₁₃ClO
MW: 136.62 g/mol
Boiling Pt: 50 °C
Density: 0.95 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00059051
CAS Number: 10503-96-5
https://us.vwr.com/stibo/web/std.lang.all/84/23/28938423.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR