TCIAC1136

C1136-1G
TCC1136-001GEA 77.03 USD
TCC1136-001G
TCIAC1136
CAS Number: 135025-34-2
MDL Number: MFCD00060060
Molecular Formula: C32H60O4
Molecular Weight: 508.83
Purity/Analysis Method: 90.0% (GC)
Form: Wax
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR