1,4-Cyclohexanedimethanolbis(2-ethylhexanoate) (cis and trans mixture) ≥98.0%
Supplier: TCI America
C1133-25G
TCC1133-025GEA
252
USD
TCC1133-025G
1,4-Cyclohexanedimethanolbis(2-ethylhexanoate) (cis and trans mixture) ≥98.0%
1,4-Cyclohexanedimethanolbis(2-ethylhexanoate) (cis and trans mixture)
CAS Number: 53148-32-6
MDL Number: MFCD00060063
Molecular Formula: C24H44O4
Molecular Weight: 396.61
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 0.95
Formula:
C₂₄H₄₄O₄ Density: 0.95 (20 °C) |
MDL Number:
MFCD00060063 CAS Number: 53148-32-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...