TCIAC1131

C1131-10G
TCC1131-010GEA 38.96 USD
TCC1131-010G
TCIAC1131
CAS Number: 65853-65-8
MDL Number: MFCD00010743
Molecular Formula: C12H10ClNO5
Molecular Weight: 283.66
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 104
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: 2-Chlorobenzyl Succinimidyl Carbonate, N-(2-Chlorocarbobenzoxyoxy)succinimide
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR