(±)-2-Chloropropionyl chloride ≥95.0% (by GC, titration analysis)
Supplier: TCI America
C1114-500G
C1114-100G
C1114-25G
TCC1114-100GEA
114.66
USD
TCC1114-100G
TCC1114-025G
TCC1114-500G
(±)-2-Chloropropionyl chloride ≥95.0% (by GC, titration analysis)
(±)-2-Chloropropionyl chloride
CAS Number: 7623-09-8
MDL Number: MFCD00000862
Molecular Formula: C3H4Cl2O
Molecular Weight: 126.96
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 111
Flash Point (°C): 31
Specific Gravity (20/20): 1.28
Formula:
C₃H₄Cl₂O Boiling Pt: 111 °C Density: 1.28 (20 °C) Flash Pt: 31 °C |
MDL Number:
MFCD00000862 CAS Number: 7623-09-8 EINECS: 231-540-7 UN: 2920 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...