1-(3-Chloropropyl)theobromine ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-(3-Chloropropyl)-3,7-dihydro-3,7-dimethyl-1H-purine-2,6-dione
CAS Number: 74409-52-2
MDL Number: MFCD00059926
Molecular Formula: C10H13ClN4O2
Molecular Weight: 256.69
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 145
Product Categories: Bioscience, Purine Alkaloids
Other Product Names / Synonyms: 1-(3-Chloropropyl)-3,7-dihydro-3,7-dimethyl-1H-purine-2,6-dione, 1-(3-Chloropropyl)-3,7-dimethylxanthine
Formula:
C₁₀H₁₃ClN₄O₂ MW: 256.69 g/mol Melting Pt: 145 °C |
MDL Number:
MFCD00059926 CAS Number: 74409-52-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...