2-(2-Chlorophenoxy)propionic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI America
C1096-25G
TCC1096-025GEA
209
USD
TCC1096-025G
2-(2-Chlorophenoxy)propionic acid ≥99.0% (by titrimetric analysis)
2-(2-Chlorophenoxy)propionic acid
CAS Number: 25140-86-7
MDL Number: MFCD00002644
Molecular Formula: C9H9ClO3
Molecular Weight: 200.62
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 116
Formula:
C₉H₉ClO₃ Melting Pt: 116 °C |
MDL Number:
MFCD00002644 CAS Number: 25140-86-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...