Cyclopropyl-2-thienyl ketone ≥96.0%

Supplier: TCI America

C1088-10G
TCC1088-010GEA 73.05 USD
TCC1088-010G
Cyclopropyl-2-thienyl ketone ≥96.0%
Cyclopropyl-2-thienyl ketone

CAS Number: 6193-47-1
MDL Number: MFCD00005427
Molecular Formula: C8H8OS
Molecular Weight: 152.21
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 92
Specific Gravity (20/20): 1.18

Formula: C₈H₈OS
Boiling Pt: 120 °C
Density: 1.18 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00005427
CAS Number: 6193-47-1
https://us.vwr.com/stibo/web/std.lang.all/83/49/28938349.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR