Ethyl cyclopropanecarboxylate ≥98.0%

Supplier: TCI America

C1060-25ML
TCC1060-025MLEA 47.6 USD
TCC1060-025ML
Ethyl cyclopropanecarboxylate ≥98.0%
Ethyl cyclopropanecarboxylate

CAS Number: 4606-07-9
MDL Number: MFCD00001282
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 18
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Cyclopropanecarboxylic Acid Ethyl Ester

Formula: C₆H₁₀O₂
MW: 114.15 g/mol
Boiling Pt: 130 °C
Density: 0.96 (20 °C)
Flash Pt: 18 °C
UN: 3272
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/83/22/28938322.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR