Hexadecyl chloroformate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Hexadecyl carbonochloridate
C1053-25ML
TCC1053-025MLEA
83.2
USD
TCC1053-025ML
Hexadecyl chloroformate ≥98.0% (by GC, titration analysis)
Hexadecyl chloroformate
CAS Number: 26272-90-2
MDL Number: MFCD00035954
Molecular Formula: C17H33ClO2
Molecular Weight: 304.90
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 110
Freezing point (°C): 14
Specific Gravity (20/20): 0.93
Other Product Names / Synonyms: Cetyl Chloroformate, Chloroformic Acid Cetyl Ester, Chloroformic Acid Hexadecyl Ester
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...