Chloropentafluoroacetone monohydrate ≥95.0% (by titrimetric analysis)

C0993-5G C0993-1G
TCC0993-005GEA 484.1 USD
TCC0993-005G TCC0993-001G
Chloropentafluoroacetone monohydrate ≥95.0% (by titrimetric analysis)
Chloropentafluoroacetone monohydrate

CAS Number: 6984-99-2
MDL Number: MFCD00150647
Molecular Formula: C3ClF5O
Molecular Weight: 182.47
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Freezing point (°C): 19
Specific Gravity (20/20): 1.73

Formula: C₃ClF₅O
MW: 200.49 g/mol
Density: 1.73 (20 °C)
MDL Number: MFCD00150647
CAS Number: 6984-99-2
https://us.vwr.com/stibo/web/std.lang.all/82/57/28938257.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR