Crocein orange G

Supplier: TCI America

Synonyms: Ponceau 4GSodium 6-hydroxy-5-(phenyldiazenyl)naphthalene-2-sulfonateacid Orange 12Sodium 6-hydroxy-5-(phenyldiazenyl)naphthalene-2-sulphonate

C0951-25G
TCC0951-025GEA 94.36 USD
TCC0951-025G
Crocein orange G
Crocein orange G

CAS Number: 1934-20-9
MDL Number: MFCD00003912
Molecular Formula: C16H12N2O4S
Molecular Weight: 350.32
Form: Crystal
Color: Yellow Red
Flash Point (°C): 93
Lambda max.: 484 nm (H2O)


Other Product Names / Synonyms: Ponceau 4G, Acid Orange 12

Formula: C₁₆H₁₂N₂O₄S
MW: 350.32 g/mol
Flash Pt: 93 °C
MDL Number: MFCD00003912
CAS Number: 1934-20-9
https://us.vwr.com/stibo/web/std.lang.all/81/91/28938191.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR