cis-3-Hexen-1-ylhexanoate ≥98.0%
Supplier: TCI America
Synonyms:
Hexanoic Acid cis-3-Hexen-1-yl Ester
C0917-25ML
TCC0917-025MLEA
30.33
USD
TCC0917-025ML
cis-3-Hexen-1-ylhexanoate ≥98.0%
cis-3-Hexen-1-ylhexanoate
CAS Number: 31501-11-8
MDL Number: MFCD00036552
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 116
Specific Gravity (20/20): 0.88
Other Product Names / Synonyms: Hexanoic Acid cis-3-Hexen-1-yl Ester
Formula:
C₁₂H₂₂O₂ MW: 198.31 g/mol Density: 0.88 (20 °C) Flash Pt: 116 °C |
MDL Number:
MFCD00036552 CAS Number: 31501-11-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...