5-Cyclohexadecen-1-one ≥97.0%
Supplier: TCI America
C0874-25G
C0874-5G
TCC0874-5GEA
32.46
USD
TCC0874-5G
TCC0874-025G
5-Cyclohexadecen-1-one ≥97.0%
5-Cyclohexadecen-1-one
CAS Number: 37609-25-9
MDL Number: MFCD00059511
Molecular Formula: C16H28O
Molecular Weight: 236.40
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 120
Flash Point (°C): 167
Freezing point (°C): 25
Formula:
C₁₆H₂₈O Boiling Pt: 120 °C Flash Pt: 167 °C |
MDL Number:
MFCD00059511 CAS Number: 37609-25-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...