N-(2-Cyanoethyl)-N-butylaniline ≥98.0%

Supplier: TCI America

Synonyms: 3-(N-Butyl-N-phenylamino)propionitrile3-(Butyl(phenyl)amino)propanenitrileN-Butyl-N-(2-cyanoethyl)aniline

C0871-25ML
TCC0871-025MLEA 33.48 USD
TCC0871-025ML
N-(2-Cyanoethyl)-N-butylaniline ≥98.0%
N-(2-Cyanoethyl)-N-butylaniline

CAS Number: 61852-40-2
MDL Number: MFCD00019863
Molecular Formula: C13H18N2
Molecular Weight: 202.30
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: N-Butyl-N-(2-cyanoethyl)aniline, 3-(N-Butyl-N-phenylamino)propionitrile

Formula: C₁₃H₁₈N₂
MW: 202.30 g/mol
Boiling Pt: 148 °C
Density: 0.99 (20 °C)
MDL Number: MFCD00019863
CAS Number: 61852-40-2
https://us.vwr.com/stibo/web/std.lang.all/80/66/28938066.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR