(3-Chloropropyl)trichlorosilane ≥97.0%

Supplier: TCI America

Synonyms: Trichloro(3-chloropropyl)silane

C0844-25G
TCC0844-025GEA 79.14 USD
TCC0844-025G
(3-Chloropropyl)trichlorosilane ≥97.0%
(3-Chloropropyl)trichlorosilane

CAS Number: 2550-06-3
MDL Number: MFCD00013694
Molecular Formula: C3H6Cl4Si
Molecular Weight: 211.97
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Flash Point (°C): 84
Specific Gravity (20/20): 1.36


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Trichloro-3-chloropropylsilane

Formula: ClH₂CCH₂CH₂SiCl₃
MW: 211.98 g/mol
Boiling Pt: 178 °C
Density: 1.36 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00013694
CAS Number: 2550-06-3
EINECS: 219-844-8
UN: 2987
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/79/84/28937984.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR