Methyl-3-cyanopropionate ≥99.0%

Supplier: TCI America

Synonyms: Methyl 3-Cyanopropionate3-Cyanopropionic Acid Methyl Ester

C0792-25ML
TCC0792-025MLEA 508 USD
TCC0792-025ML
Methyl-3-cyanopropionate ≥99.0%
Methyl-3-cyanopropionate

CAS Number: 4107-62-4
MDL Number: MFCD00058965
Molecular Formula: C5H7NO2
Molecular Weight: 113.12
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 3-Cyanopropionic Acid Methyl Ester

Formula: C₅H₇NO₂
MW: 113.12 g/mol
Boiling Pt: 216 °C
Density: 1.08 (20 °C)
MDL Number: MFCD00058965
CAS Number: 4107-62-4
https://us.vwr.com/stibo/web/std.lang.all/78/29/28937829.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR