Azelaic dihydrazide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Nonanedihydrazide

A0746-25G
TCA0746-025GEA 139.01 USD
TCA0746-025G
Azelaic dihydrazide ≥95.0% (by titrimetric analysis)
Azelaic dihydrazide

CAS Number: 4080-95-9
MDL Number: MFCD00051529
Molecular Formula: C9H20N4O2
Molecular Weight: 216.29
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 183

Formula: C₉H₂₀N₄O₂
MW: 216.29 g/mol
Melting Pt: 183 °C
MDL Number: MFCD00051529
CAS Number: 4080-95-9
https://us.vwr.com/stibo/web/std.lang.all/84/56/28928456.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR