2-Chlorophenylhydrazine sulfate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

C0754-25G
TCC0754-025GEA 140.01 USD
TCC0754-025G
2-Chlorophenylhydrazine sulfate ≥95.0% (by titrimetric analysis)
2-Chlorophenylhydrazine sulfate

MDL Number: MFCD00060255
Molecular Formula: C6H7ClN2
Molecular Weight: 383.24
Purity/Analysis Method: 95.0% (T)
Form: Crystal

Formula: C₁₂H₁₄Cl₂N₄.H₂SO₄
MW: 383.24 g/mol
MDL Number: MFCD00060255
CAS Number: 198279-94-6
https://us.vwr.com/stibo/web/std.lang.all/77/97/28937797.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR