Octyl chloroformate ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C0724-25ML
TCC0724-025MLEA 176.25 USD
TCC0724-025ML
Octyl chloroformate ≥97.0% (by GC, titration analysis)
Octyl chloroformate

CAS Number: 7452-59-7
MDL Number: MFCD00000652
Molecular Formula: C9H17ClO2
Molecular Weight: 192.68
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 76
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Chloroformic Acid n-Octyl Ester

https://us.vwr.com/stibo/web/std.lang.all/77/70/28937770.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR