Heptyl chloroformate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C0722-25ML
TCC0722-025MLEA 115 USD
TCC0722-025ML
Heptyl chloroformate ≥98.0% (by GC, titration analysis)
Heptyl chloroformate

CAS Number: 33758-34-8
MDL Number: MFCD00059484
Molecular Formula: C8H15ClO2
Molecular Weight: 178.66
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 71
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Chloroformic Acid Heptyl Ester

https://us.vwr.com/stibo/web/std.lang.all/77/68/28937768.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR