4'-Chloroacetoacetanilide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-(4-Chlorophenyl)-3-oxobutanamide

C0625-25G
TCC0625-025GEA 46 USD
TCC0625-025G
4'-Chloroacetoacetanilide ≥98.0% (by titrimetric analysis)
4'-Chloroacetoacetanilide

CAS Number: 101-92-8
MDL Number: MFCD00000613
Molecular Formula: C10H10ClNO2
Molecular Weight: 211.65
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 132
Flash Point (°C): 176


Other Product Names / Synonyms: N-Acetoacetyl-4-chloroaniline

https://us.vwr.com/stibo/web/std.lang.all/76/62/28937662.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR