2-Chloroethyl methyl sulfide ≥98.0%
Supplier: TCI America
C0588-10G
TCC0588-010GEA
87
USD
TCC0588-010G
2-Chloroethyl methyl sulfide ≥98.0%
2-Chloroethyl methyl sulphide
CAS Number: 542-81-4
MDL Number: MFCD00000978
Molecular Formula: C3H7ClS
Molecular Weight: 110.60
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 42
Specific Gravity (20/20): 1.13
Storage Temperature: 0-10°C
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...