Coriphosphine O

Supplier: TCI America

Synonyms: Basic Yellow 7

C0539-1G
TCC0539-1GEA 101.47 USD
TCC0539-1G
Coriphosphine O
Coriphosphine O

CAS Number: 5409-37-0
MDL Number: MFCD00050595
Molecular Formula: C16H18ClN3
Molecular Weight: 287.79
Form: Crystal
Color: Yellow Red


Other Product Names / Synonyms: Basic Yellow 7

Formula: C₁₆H₁₈ClN₃
MW: 287.79 g/mol
MDL Number: MFCD00050595
CAS Number: 5409-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR