Ethyl-ɑ-cyanocinnamate ≥98.0%

Supplier: TCI America

Synonyms: 2-Cyano-3-phenylacrylic Acid Ethyl Esteralpha-Cyanocinnamic Acid Ethyl EsterEthyl 2-Cyano-3-phenylacrylate

C0448-25G
TCC0448-025GEA 135.96 USD
TCC0448-025G
Ethyl-ɑ-cyanocinnamate ≥98.0%
Ethyl-alpha-cyanocinnamate

CAS Number: 2025-40-3
MDL Number: MFCD00009137
Molecular Formula: C12H11NO2
Molecular Weight: 201.23
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 188
Melting point (°C): 50


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 2-Cyano-3-phenylacrylic Acid Ethyl Ester, alpha-Cyanocinnamic Acid Ethyl Ester, Ethyl 2-Cyano-3-phenylacrylate

Formula: C₁₂H₁₁NO₂
MW: 201.23 g/mol
Boiling Pt: 188 °C
Melting Pt: 50 °C
MDL Number: MFCD00009137
CAS Number: 2025-40-3
https://us.vwr.com/stibo/web/std.lang.all/74/72/28937472.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR