TOKYC0422

C0422-25ML
TCC0422-025MLEA 26.02 USD
TCC0422-025ML
TOKYC0422
CAS Number: 4786-20-3
MDL Number: MFCD00001935
Molecular Formula: C4H5N
Molecular Weight: 67.09
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 16
Specific Gravity (20/20): 0.83

Other Product Names / Synonyms: 2-Butenenitrile
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR