Crotonamide ≥98.0%

Supplier: TCI America

Synonyms: trans-2-Butenamide

C0415-25G
TCC0415-025GEA 165.37 USD
TCC0415-025G
Crotonamide ≥98.0%
Crotonamide

CAS Number: 23350-58-5
MDL Number: MFCD00059859
Molecular Formula: C4H7NO
Molecular Weight: 85.11
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 158


Other Product Names / Synonyms: trans-2-Butenamide

Formula: C₄H₇NO
MW: 85.11 g/mol
Melting Pt: 158 °C
MDL Number: MFCD00059859
CAS Number: 23350-58-5
https://us.vwr.com/stibo/web/std.lang.all/74/44/28937444.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR