Cortisone acetate ≥99.0% (by HPLC)

Synonyms: Cortisone 21-acetate

C0389-1G C0389-25G C0389-5G
TCC0389-25GEA 460 USD
TCC0389-25G TCC0389-005G TCC0389-001G
Cortisone acetate ≥99.0% (by HPLC)
Cortisone acetate

CAS Number: 50-04-4
MDL Number: MFCD00003609
Molecular Formula: C23H30O6
Molecular Weight: 402.49
Purity/Analysis Method: 99.0% (HPLC)
Form: Crystal
Melting point (°C): 238
Specific rotation [a]20/D: 212 deg (C=1, MeOH)

Formula: C₂₃H₃₀O₆
MW: 402.49 g/mol
Melting Pt: 238 °C
MDL Number: MFCD00003609
CAS Number: 50-04-4
https://us.vwr.com/stibo/web/std.lang.all/74/14/28937414.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR