Chrome pure blue BX
Supplier: TCI America
Synonyms:
Sunchromine Pure Blue BX
, Mordant Blue 1
, Eriochrome Azurol B
C0337-25G
TCC0337-025GEA
103.86
USD
TCC0337-025G
Chrome pure blue BX
Chrome pure blue BX
CAS Number: 1796-92-5
MDL Number: MFCD00036446
Molecular Formula: C23H16Cl2O6
Molecular Weight: 503.24
Form: Crystal
Color: Reddish Yellow
Other Product Names / Synonyms: Eriochrome Azurol B, Sunchromine Pure Blue BX, Mordant Blue 1
Formula:
C₂₃H₁₆Cl₂O₆ MW: 503.24 g/mol |
MDL Number:
MFCD00036446 CAS Number: 1796-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...