2-Chloro-4-phenylphenol ≥96.0%

Supplier: TCI America

Synonyms: 3-Chlorobiphenyl-4-ol3-Chloro-4-hydroxybiphenyl

C0263-25G
TCC0263-025GEA 71.03 USD
TCC0263-025G
2-Chloro-4-phenylphenol ≥96.0%
2-Chloro-4-phenylphenol

CAS Number: 92-04-6
MDL Number: MFCD00045745
Molecular Formula: C12H9ClO
Molecular Weight: 204.65
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Boiling point (°C): 322
Melting point (°C): 79
Flash Point (°C): 174


Other Product Names / Synonyms: 3-Chlorobiphenyl-4-ol, 3-Chloro-4-hydroxybiphenyl

Formula: C₁₂H₉ClO
MW: 204.65 g/mol
Boiling Pt: 322 °C
Melting Pt: 79 °C
Flash Pt: 174 °C
MDL Number: MFCD00045745
CAS Number: 92-04-6
https://us.vwr.com/stibo/web/std.lang.all/73/11/28937311.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR