Erio green B
Supplier: TCI America
Synonyms:
Pontacyl Green NV
, Lissamine Green V
, Acid Brilliant Green J
, Acid Green 16
A0687-5G
TCA0687-005GEA
117.71
USD
TCA0687-005G
Erio green B
Erio green B
CAS Number: 12768-78-4
MDL Number: MFCD00060075
Molecular Formula: C31H34N2O6S2
Molecular Weight: 616.72
Form: Crystal
Color: Red
Other Product Names / Synonyms: Acid Brilliant Green J, Acid Green 16, Lissamine Green V, Pontacyl Green NV
Formula:
C₃₁H₃₄N₂O₆S₂ MW: 616.72 g/mol |
MDL Number:
MFCD00060075 CAS Number: 12768-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...