4,4'-Azoxyphenetole ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
4,4'-Azoxydiphenetole
, 4,4'-Diethoxyazoxybenzene
A0683-1G
A0683-5G
TCA0683-5GEA
185.67
USD
TCA0683-5G
TCA0683-001G
4,4'-Azoxyphenetole ≥98.0% (by total nitrogen basis)
4,4'-Azoxyphenetole
CAS Number: 4792-83-0
MDL Number: MFCD00026790
Molecular Formula: C16H18N2O3
Molecular Weight: 286.33
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 132
Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4,4'-Diethoxyazoxybenzene
Formula:
C₁₆H₁₈N₂O₃ MW: 286.33 g/mol Melting Pt: 132 °C |
MDL Number:
MFCD00026790 CAS Number: 4792-83-0 EINECS: 225-347-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...