Butyl chloroacetate ≥98.0%

Supplier: TCI America

Synonyms: Chloroacetic Acid Butyl Ester

C0091-25ML
TCC0091-025MLEA 31.45 USD
TCC0091-025ML
Butyl chloroacetate ≥98.0%
Butyl chloroacetate

CAS Number: 590-02-3
MDL Number: MFCD00018925
Molecular Formula: C6H11ClO2
Molecular Weight: 150.60
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Chloroacetic Acid Butyl Ester

Formula: C₆H₁₁ClO₂
MW: 150.60 g/mol
Boiling Pt: 183 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00018925
CAS Number: 590-02-3
https://us.vwr.com/stibo/web/std.lang.all/71/73/28937173.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR