Chloramine-B-hydrate ≥80.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Benzenesulfonechloramide Sodium Salt

C0075-500G C0075-25G
TCC0075-500GEA 267.56 USD
TCC0075-500G TCC0075-025G
Chloramine-B-hydrate ≥80.0% (by titrimetric analysis)
Chloramine-B-hydrate

CAS Number: 127-52-6
MDL Number: MFCD00007545
Molecular Formula: C6H6ClNO2S
Molecular Weight: 213.61
Purity/Analysis Method: 80.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: Benzenesulfonechloramide Sodium Salt

Formula: C₆H₆ClNO₂S
MW: 213.61 g/mol
MDL Number: MFCD00007545
CAS Number: 127-52-6
https://us.vwr.com/stibo/web/std.lang.all/71/59/28937159.jpg

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR