Cellulose TEAE Capacity: 0.72 meq/g

Synonyms: Triethylaminoethyl Cellulose

C0068-10G
TCC0068-010GEA 67.32 USD
TCC0068-010G
Cellulose TEAE Capacity: 0.72 meq/g
Cellulose TEAE

MDL Number: MFCD00146355
Form: Crystal
Color: White


Other Product Names / Synonyms: Triethylaminoethyl Cellulose

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR