Bis(2-chloroethyl)carbonate ≥98.0%

Supplier: TCI America

Synonyms: Carbonic Acid Bis(2-chloroethyl) EsterCarbonic Acid Di(2-chloroethyl) EsterDi(2-chloroethyl) Carbonate

C0038-10G
TCC0038-010GEA 189.73 USD
TCC0038-010G
Bis(2-chloroethyl)carbonate ≥98.0%
Bis(2-chloroethyl)carbonate

CAS Number: 623-97-2
MDL Number: MFCD00018962
Molecular Formula: C5H8Cl2O3
Molecular Weight: 187.02
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 241
Freezing point (°C): 10
Specific Gravity (20/20): 1.35


Other Product Names / Synonyms: Carbonic Acid Bis(2-chloroethyl) Ester, Carbonic Acid Di(2-chloroethyl) Ester, Di(2-chloroethyl) Carbonate

Formula: C₅H₈Cl₂O₃
MW: 187.02 g/mol
Boiling Pt: 241 °C
Density: 1.35 (20 °C)
MDL Number: MFCD00018962
CAS Number: 623-97-2
https://us.vwr.com/stibo/web/std.lang.all/71/40/28937140.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR