(±)-Camphene ≥78.0% (contains ca. 20% Tricyclene)
Supplier: TCI America
Synonyms:
(±)-3,3-Dimethyl-2-methylennorcamphane
C0009-25G
TCC0009-025GEA
16.24
USD
TCC0009-025G
(±)-Camphene ≥78.0% (contains ca. 20% Tricyclene)
(±)-Camphene
CAS Number: 79-92-5
MDL Number: MFCD00066603
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 46
Flash Point (°C): 33
Formula:
C₁₀H₁₆ MW: 136.24 g/mol Boiling Pt: 159 °C Melting Pt: 46 °C Density: 0.8421 g/cm³ (20 °C) Flash Pt: 33 °C |
MDL Number:
MFCD00066603 CAS Number: 79-92-5 EINECS: 201-234-8 UN: 1325 ADR: 4.1,III Merck Index: 12,01777 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...