Cacotheline Monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0001-5G C0001-1G
TCC0001-005GEA 258.78 USD
TCC0001-005G TCC0001-1G
Cacotheline Monohydrate ≥98.0% (by titrimetric analysis)
Cacotheline monohydrate

CAS Number: 561-20-6
MDL Number: MFCD00077211
Molecular Formula: C21H21N3O7
Molecular Weight: 490.43
Purity/Analysis Method: 98.0% (T)
Form: Crystal

Formula: C₂₁H₂₁N₃O₇
MDL Number: MFCD00077211
CAS Number: 561-20-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR