Bis(zinc porphyrin) ∼5 µmol/L in dichloromethane
B3515-5ML
TCB3515-5MLEA
28.56
USD
TCB3515-5ML
Bis(zinc porphyrin) ∼5 µmol/L in dichloromethane
Bis(zinc porphyrin)
[for CD Spectroscopy]
CAS Number: 92995-45-4
MDL Number: MFCD03459124
Molecular Formula: C74H90N8Zn2
Molecular Weight: 1222.35
Form: Clear Liquid
Color: Yellow
Storage Temperature: 0-10°C
Formula:
C₇₄H₉₀N₈Zn₂ |
MDL Number:
MFCD03459124 CAS Number: 92995-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...