1-Acetyl-3-methylurea ≥98.0% (by total nitrogen basis)
Supplier: TCI America
A0101-25G
TCA0101-025GEA
148.13
USD
TCA0101-025G
1-Acetyl-3-methylurea ≥98.0% (by total nitrogen basis)
1-Acetyl-3-methylurea
CAS Number: 623-59-6
MDL Number: MFCD00026167
Molecular Formula: C4H8N2O2
Molecular Weight: 116.12
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 180
Formula:
C₄H₈N₂O₂ Melting Pt: 180 °C |
MDL Number:
MFCD00026167 CAS Number: 623-59-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...