1-Bromo-3-(trimethylsilyl)benzene ≥96.0%

Supplier: TCI America

Synonyms: (3-BRomophenyl)trimethylsilane

B3477-5G
TCB3477-5GEA 143 USD
TCB3477-5G
1-Bromo-3-(trimethylsilyl)benzene ≥96.0%
1-Bromo-3-(trimethylsilyl)benzene

CAS Number: 17878-47-6
MDL Number: MFCD00093882
Molecular Formula: C9H13BrSi
Molecular Weight: 229.19
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 50
Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: (3-Bromophenyl)trimethylsilane

Formula: C₉H₁₃BrSi
MW: 229.1890 g/mol
Boiling Pt: 50 °C
Density: 1.22 (20 °C)
MDL Number: MFCD00093882
CAS Number: 17878-47-6

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR