8-Bromotheophylline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3456-1G B3456-5G
TCB3456-1GEA 165 USD
TCB3456-1G TCB3456-5G
8-Bromotheophylline ≥98.0% (by HPLC, titration analysis)
8-Bromotheophylline

CAS Number: 10381-75-6
MDL Number: MFCD00022664
Molecular Formula: C7H7BrN4O2
Molecular Weight: 259.06
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 295

Formula: C₇H₇BrN₄O₂
Melting Pt: 295 °C
MDL Number: MFCD00022664
CAS Number: 10381-75-6
https://us.vwr.com/stibo/web/std.lang.all/48/17/28934817.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR