Bis[2-(trimethylsilyloxy)ethyleEther ≥98.0%

Synonyms: Diethylene Glycol Bis(trimethylsilyl Ether)

B3378-25G B3378-5G
TCB3378-25GEA 199.82 USD
TCB3378-25G TCB3378-5G
Bis[2-(trimethylsilyloxy)ethyleEther ≥98.0%
Bis[2-(trimethylsilyloxy)ethyleEther

CAS Number: 16654-74-3
Molecular Formula: C10H26O3Si2
Molecular Weight: 250.49
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 87
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Diethylene Glycol Bis(trimethylsilyl Ether)

Formula: C₁₀H₂₆O₃Si₂
MW: 250.49 g/mol
Boiling Pt: 87 °C
Density: 0.89 (20 °C)
CAS Number: 16654-74-3
https://us.vwr.com/stibo/web/std.lang.all/47/61/28934761.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR