5,5'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,2'-Bithiophene-5,5'-diboronic Acid Bis(pinacol) Ester

B3363-5G
TCB3363-5GEA 308.47 USD
TCB3363-5G
5,5'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene ≥98.0% (by titrimetric analysis)
5,5'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene

CAS Number: 239075-02-6
MDL Number: MFCD06798112
Molecular Formula: C20H28B2O4S2
Molecular Weight: 418.18
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 207


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Semiconducting Polymers, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2,2'-Bithiophene-5,5'-diboronic Acid Bis(pinacol) Ester

Formula: C₂₀H₂₈B₂O₄S₂
MW: 418.18 g/mol
Melting Pt: 207 °C
MDL Number: MFCD06798112
CAS Number: 239075-02-6
https://us.vwr.com/stibo/web/std.lang.all/47/50/28934750.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR