TCIAB3321

B3321-25G
TCB3321-25GEA 37 USD
TCB3321-25G
TCIAB3321
CAS Number: 4437-85-8
MDL Number: MFCD00143317
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 74
Flash Point (°C): 135
Specific Gravity (20/20): 1.15

Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 4-Ethyl-1,3-dioxolan-2-one
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR