Benzyl-4-fluorophenyl ketone ≥98.0%

Supplier: TCI America

Synonyms: 1-(4-Fluorophenyl)-2-phenylethanone

B3213-25G
TCB3213-25GEA 131.95 USD
TCB3213-25G
Benzyl-4-fluorophenyl ketone ≥98.0%
Benzyl 4-fluorophenyl ketone

CAS Number: 347-84-2
MDL Number: MFCD00017960
Molecular Formula: C14H11FO
Molecular Weight: 214.24
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 82


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4'-Fluoro-2-phenylacetophenone

Formula: C₁₄H₁₁FO
MW: 214.2350 g/mol
Melting Pt: 82 °C
MDL Number: MFCD00017960
CAS Number: 347-84-2
https://us.vwr.com/stibo/web/std.lang.all/46/36/28934636.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR