Dibenzyl adipate ≥95.0%
Supplier: TCI America
Synonyms:
Adipic Acid Dibenzyl Ester
A0618-25G
TCA0618-025GEA
219.75
USD
TCA0618-025G
Dibenzyl adipate ≥95.0%
Dibenzyl adipate
CAS Number: 2451-84-5
MDL Number: MFCD00059667
Molecular Formula: C20H22O4
Molecular Weight: 326.39
Purity/Analysis Method:
Form: Crystal
Freezing point (°C): 36
Other Product Names / Synonyms: Adipic Acid Dibenzyl Ester
Formula:
C₂₀H₂₂O₄ MW: 326.39 g/mol |
MDL Number:
MFCD00059667 CAS Number: 2451-84-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...