Acetyl iodide ≥95.0% (by titrimetric analysis)
Supplier: TCI America
A0095-25G
A0095-10G
TCA0095-010GEA
263.82
USD
TCA0095-010G
TCA0095-25G
Acetyl iodide ≥95.0% (by titrimetric analysis)
Acetyl iodide
CAS Number: 507-02-8
MDL Number: MFCD00039399
Molecular Formula: C2H3IO
Molecular Weight: 169.95
Purity/Analysis Method:
Form: Liquid
Boiling point (°C): 108
Specific Gravity (20/20): 2.07
Formula:
C₂H₃IO Boiling Pt: 108 °C Density: 2.07 (20 °C) |
UN:
1898 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...