Bis(tri-tert-butylphosphine)platinum(0)
Supplier: TCI America
Synonyms:
Bis(tri-t-butylphosphine)platinum (0)
B3162-250MG
TCB3162-250MGEA
285
USD
TCB3162-250MG
Bis(tri-tert-butylphosphine)platinum(0)
Bis(tri-tert-butylphosphine)platinum(0)
CAS Number: 60648-70-6
MDL Number: MFCD04974248
Molecular Formula: C24H54P2Pt
Molecular Weight: 599.73
Form: Crystal
Color: White
Melting point (°C): 269
Formula:
C₂₄H₅₄P₂Pt MW: 599.73 g/mol |
MDL Number:
MFCD04974248 CAS Number: 60648-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...